3-(4-Bromophenyl)-1H-pyrazole-4-carboxylic acid

Supplier: Matrix Scientific

038661-500MG
101845-958EA 80.5 USD
101845-958
3-(4-Bromophenyl)-1H-pyrazole-4-carboxylic acid
3-(4-Bromophenyl)-1H-pyrazole-4-carboxylic acid

Hazard: Irritant

Formula: C₁₀H₇BrN₂O₂
MDL Number: MFCD03834508
CAS Number: 618383-49-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR