trans-2-Bromocinnamic acid

Supplier: Matrix Scientific

038653-500MG
101845-942EA 116.92 USD
101845-942
trans-2-Bromocinnamic acid
trans-2-Bromocinnamic acid

Hazard: Irritant

Formula: C₉H₇BrO₂
MW: 227.06 g/mol
MDL Number: MFCD00004382
CAS Number: 7499-56-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR