Ethyl-3-amino-3-(4-isopropoxyphenyl)propanoate hydrochloride

Supplier: Matrix Scientific

038650-500MG
101845-936EA 144.9 USD
101845-936
Ethyl-3-amino-3-(4-isopropoxyphenyl)propanoate hydrochloride
Ethyl-3-amino-3-(4-isopropoxyphenyl)propanoate hydrochloride

Hazard: Irritant

Formula: C₁₄H₂₂ClNO₃
MDL Number: MFCD06653254
CAS Number: 945419-73-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR