(1-Aminocyclohexyl)acetic acid

Supplier: Matrix Scientific

Synonyms: 2-(1-Aminocyclohexyl)acetic acid

038646-500MG
101845-928EA 165.6 USD
101845-928
(1-Aminocyclohexyl)acetic acid
(1-Aminocyclohexyl)acetic acid

Hazard: Irritant

Formula: C₈H₁₅NO₂
MW: 157.21 g/mol
MDL Number: MFCD02258116
CAS Number: 37631-92-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR