(2E)-3-[4-(2-Amino-2-oxoethoxy)phenyl]acrylic acid

Supplier: Matrix Scientific

038641-500MG
101845-918EA 109.25 USD
101845-918
(2E)-3-[4-(2-Amino-2-oxoethoxy)phenyl]acrylic acid
(2E)-3-[4-(2-Amino-2-oxoethoxy)phenyl]acrylic acid

Hazard: Irritant

Formula: C₁₁H₁₁NO₄
MDL Number: MFCD02256074
CAS Number: 873811-55-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR