(2E)-3-(5-Methyl-2-furyl)acrylic acid

Supplier: Matrix Scientific

038638-500MG
101845-910EA 87.4 USD
101845-910
(2E)-3-(5-Methyl-2-furyl)acrylic acid
(2E)-3-(5-Methyl-2-furyl)acrylic acid

Hazard: Irritant

Formula: C₈H₈O₃
MDL Number: MFCD00186942
CAS Number: 14779-25-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR