2-Chloro-4-oxo-4H-pyrido[1,2-a]pyrimidine-3-carbaldehyde

Supplier: Matrix Scientific

038628-500MG
101845-888EA 289.8 USD
101845-888
2-Chloro-4-oxo-4H-pyrido[1,2-a]pyrimidine-3-carbaldehyde
2-Chloro-4-oxo-4H-pyrido[1,2-a]pyrimidine-3-carbaldehyde

Hazard: Irritant

Formula: C₉H₅ClN₂O₂
MDL Number: MFCD00858192
CAS Number: 17481-62-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR