4-(5-Formyl-2-furyl)benzoic acid

Supplier: Matrix Scientific

038627-500MG
101845-886EA 186.3 USD
101845-886
4-(5-Formyl-2-furyl)benzoic acid
4-(5-Formyl-2-furyl)benzoic acid

Hazard: Irritant

Formula: C₁₂H₈O₄
MDL Number: MFCD00455087
CAS Number: 39245-15-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR