3-Methylisatoic anhydride

Supplier: Matrix Scientific

Synonyms: 8-Methyl-1H-benzo[d][1,3]oxazine-2,4-dione

038625-500MG
101845-882EA 290.77 USD
101845-882
3-Methylisatoic anhydride
3-Methylisatoic anhydride

Hazard: Irritant

Formula: C₉H₇NO₃
MW: 177.1568 g/mol
MDL Number: MFCD03426399
CAS Number: 66176-17-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR