1-(2-Methoxyethyl)-5-(4-methoxyphenyl)-2,4-dimethyl-1H-pyrrole-3-carboxylic acid

Supplier: Matrix Scientific

038614-500MG
101845-860EA 217.35 USD
101845-860
1-(2-Methoxyethyl)-5-(4-methoxyphenyl)-2,4-dimethyl-1H-pyrrole-3-carboxylic acid
1-(2-Methoxyethyl)-5-(4-methoxyphenyl)-2,4-dimethyl-1H-pyrrole-3-carboxylic acid

Hazard: Irritant

Formula: C₁₇H₂₁NO₄
MDL Number: MFCD12027408
CAS Number: 879329-83-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR