Ethyl-2-hydrazino-4-methyl-1,3-thiazole-5-carboxylate

Supplier: Matrix Scientific

038607-500MG
101845-846EA 241.5 USD
101845-846
Ethyl-2-hydrazino-4-methyl-1,3-thiazole-5-carboxylate
Ethyl-2-hydrazino-4-methyl-1,3-thiazole-5-carboxylate

Hazard: Irritant

Formula: C₇H₁₁N₃O₂S
MDL Number: MFCD01859678
CAS Number: 52481-66-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR