3-Methyl-2-(1-oxo-1,3-dihydro-2H-isoindol-2-yl)pentanoic acid

Supplier: Matrix Scientific

038605-500MG
101845-842EA 109.25 USD
101845-842
3-Methyl-2-(1-oxo-1,3-dihydro-2H-isoindol-2-yl)pentanoic acid
3-Methyl-2-(1-oxo-1,3-dihydro-2H-isoindol-2-yl)pentanoic acid

Hazard: Irritant

Formula: C₁₄H₁₇NO₃
MDL Number: MFCD04367477
CAS Number: 180923-81-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR