4-[5-(4-Methoxyphenyl)-1H-pyrazol-3-yl]butanoic acid

Supplier: Matrix Scientific

038602-500MG
101845-836EA 217.35 USD
101845-836
4-[5-(4-Methoxyphenyl)-1H-pyrazol-3-yl]butanoic acid
4-[5-(4-Methoxyphenyl)-1H-pyrazol-3-yl]butanoic acid

Hazard: Irritant

Formula: C₁₄H₁₆N₂O₃
MDL Number: MFCD12027404
CAS Number: 1239069-86-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR