4-(6-Chloropyridazin-3-yl)morpholine

Supplier: Matrix Scientific

038600-500MG
101845-832EA 217.35 USD
101845-832
4-(6-Chloropyridazin-3-yl)morpholine
4-(6-Chloropyridazin-3-yl)morpholine

Hazard: Irritant

Formula: C₈H₁₀ClN₃O
MW: 199.6375 g/mol
MDL Number: MFCD00574575
CAS Number: 17259-32-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR