1-(1-Adamantyl)propan-2-amine

Supplier: Matrix Scientific

038598-500MG
101845-828EA 87.4 USD
101845-828
1-(1-Adamantyl)propan-2-amine
1-(1-Adamantyl)propan-2-amine

Hazard: Irritant

Formula: C₁₃H₂₃N
MDL Number: MFCD02632259
CAS Number: 39978-69-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR