4-(1-Adamantyl)-4-aminobutanoic acid

Supplier: Matrix Scientific

038592-500MG
101845-816EA 241.5 USD
101845-816
4-(1-Adamantyl)-4-aminobutanoic acid
4-(1-Adamantyl)-4-aminobutanoic acid

Hazard: Irritant

Formula: C₁₄H₂₃NO₂
MDL Number: MFCD09035621
CAS Number: 933721-54-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR