1-(1-Adamantyl)pentane-1,4-dione

Supplier: Matrix Scientific

038589-500MG
101845-808EA 289.8 USD
101845-808
1-(1-Adamantyl)pentane-1,4-dione
1-(1-Adamantyl)pentane-1,4-dione

Hazard: Irritant

Formula: C₁₅H₂₂O₂
MDL Number: MFCD09035444
CAS Number: 195137-34-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR