[5-(3,4-Dimethoxyphenyl)-4-oxothieno-[2,3-d]pyrimidin-3(4H)-yl]acetic acid

Supplier: Matrix Scientific

038582-500MG
101845-794EA 217.35 USD
101845-794
[5-(3,4-Dimethoxyphenyl)-4-oxothieno-[2,3-d]pyrimidin-3(4H)-yl]acetic acid
[5-(3,4-Dimethoxyphenyl)-4-oxothieno-[2,3-d]pyrimidin-3(4H)-yl]acetic acid

Hazard: Irritant

Formula: C₁₆H₁₄N₂O₅S
MDL Number: MFCD03016034
CAS Number: 439140-53-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR