2-Morpholin-4-ylethyl chloroacetate

Supplier: Matrix Scientific

038576-500MG
101845-782EA 144.9 USD
101845-782
2-Morpholin-4-ylethyl chloroacetate
2-Morpholin-4-ylethyl chloroacetate

Hazard: Irritant

Formula: C₈H₁₄ClNO₃
MDL Number: MFCD03856577
CAS Number: 626210-61-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR