Octahydro-2H-benzimidazole-2-thione

Supplier: Matrix Scientific

Synonyms: Hexahydro-1H-benzo[d]imidazole-2(3H)-thione

038573-500MG
101845-776EA 144.9 USD
101845-776
Octahydro-2H-benzimidazole-2-thione
Octahydro-2H-benzimidazole-2-thione

Hazard: Irritant

Formula: C₇H₁₂N₂S
MW: 156.25 g/mol
MDL Number: MFCD00123649
CAS Number: 30086-64-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR