2-Hydroxy-7-methylquinoline-3-carboxylic acid

Supplier: Matrix Scientific

038567-500MG
101845-764EA 250.7 USD
101845-764
2-Hydroxy-7-methylquinoline-3-carboxylic acid
2-Hydroxy-7-methylquinoline-3-carboxylic acid

Hazard: Irritant

Formula: C₁₁H₉NO₃
MDL Number: MFCD12027396
CAS Number: 436087-28-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR