1-(3,5-Dimethylphenyl)-1H-benzimidazole-5-carboxylic acid

Supplier: Matrix Scientific

038561-500MG
101845-752EA 217.35 USD
101845-752
1-(3,5-Dimethylphenyl)-1H-benzimidazole-5-carboxylic acid
1-(3,5-Dimethylphenyl)-1H-benzimidazole-5-carboxylic acid

Hazard: Irritant

Formula: C₁₆H₁₄N₂O₂
MDL Number: MFCD03021469
CAS Number: 442531-63-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR