1-[4-(Benzyloxy)phenyl]-2-methyl-1H-benzimidazole-5-carboxylic acid
Supplier: Matrix Scientific
038554-500MG
101845-738EA
217.35
USD
101845-738
1-[4-(Benzyloxy)phenyl]-2-methyl-1H-benzimidazole-5-carboxylic acid
1-[4-(Benzyloxy)phenyl]-2-methyl-1H-benzimidazole-5-carboxylic acid
Hazard: Irritant
Formula:
C₂₂H₁₈N₂O₃ |
MDL Number:
MFCD02939958 CAS Number: 452088-73-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...