1-(4-Methoxyphenyl)-2-methyl-1H-benzimidazole-5-carboxylic acid

Supplier: Matrix Scientific

038548-500MG
101845-726EA 217.35 USD
101845-726
1-(4-Methoxyphenyl)-2-methyl-1H-benzimidazole-5-carboxylic acid
1-(4-Methoxyphenyl)-2-methyl-1H-benzimidazole-5-carboxylic acid

Hazard: Irritant

Formula: C₁₆H₁₄N₂O₃
MDL Number: MFCD02939955
CAS Number: 442531-47-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR