1-Ethyl-4-(piperidin-4-ylmethyl)piperazine

Supplier: Matrix Scientific

038528-500MG
101845-686EA 217.35 USD
101845-686
1-Ethyl-4-(piperidin-4-ylmethyl)piperazine
1-Ethyl-4-(piperidin-4-ylmethyl)piperazine

Hazard: Irritant

Formula: C₁₂H₂₅N₃
MDL Number: MFCD05215227
CAS Number: 775288-25-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR