6-Chloro-2-methyl-3,4-dihydro-2H-1,4-benzoxazine

Supplier: Matrix Scientific

038523-500MG
101845-676EA 332.35 USD
101845-676
6-Chloro-2-methyl-3,4-dihydro-2H-1,4-benzoxazine
6-Chloro-2-methyl-3,4-dihydro-2H-1,4-benzoxazine

Hazard: Irritant

Formula: C₉H₁₀ClNO
MDL Number: MFCD11603432
CAS Number: 1082837-97-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR