N-Benzyl-2-bromopropanamide

Supplier: Matrix Scientific

038517-500MG
101845-664EA 87.4 USD
101845-664
N-Benzyl-2-bromopropanamide
N-Benzyl-2-bromopropanamide

Hazard: Irritant

Formula: C₁₀H₁₂BrNO
MDL Number: MFCD01696041
CAS Number: 6653-71-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR