2-Bromo-N-(2,4-dimethoxyphenyl)butanamide

Supplier: Matrix Scientific

038509-500MG
101845-648EA 87.4 USD
101845-648
2-Bromo-N-(2,4-dimethoxyphenyl)butanamide
2-Bromo-N-(2,4-dimethoxyphenyl)butanamide

Hazard: Irritant

Formula: C₁₂H₁₆BrNO₃
MDL Number: MFCD12027375
CAS Number: 1119451-52-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR