2-Bromo-N-(4-fluorobenzyl)propanamide

Supplier: Matrix Scientific

038506-500MG
101845-642EA 109.25 USD
101845-642
2-Bromo-N-(4-fluorobenzyl)propanamide
2-Bromo-N-(4-fluorobenzyl)propanamide

Hazard: Irritant

Formula: C₁₀H₁₁BrFNO
MDL Number: MFCD12027372
CAS Number: 1119451-51-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR