2-Bromo-N-(5-chloro-2,4-dimethoxyphenyl)propanamide

Supplier: Matrix Scientific

038505-500MG
101845-640EA 87.4 USD
101845-640
2-Bromo-N-(5-chloro-2,4-dimethoxyphenyl)propanamide
2-Bromo-N-(5-chloro-2,4-dimethoxyphenyl)propanamide

Hazard: Irritant

Formula: C₁₁H₁₃BrClNO₃
MDL Number: MFCD12027371
CAS Number: 1119452-45-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR