2-Bromo-N-[2-(4-chlorophenyl)ethyl]propanamide

Supplier: Matrix Scientific

038501-500MG
101845-632EA 109.25 USD
101845-632
2-Bromo-N-[2-(4-chlorophenyl)ethyl]propanamide
2-Bromo-N-[2-(4-chlorophenyl)ethyl]propanamide

Hazard: Irritant

Formula: C₁₁H₁₃BrClNO
MDL Number: MFCD12027368
CAS Number: 1119450-46-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR