2-Bromo-N-(2,5-difluorophenyl)butanamide

Supplier: Matrix Scientific

038499-500MG
101845-628EA 109.25 USD
101845-628
2-Bromo-N-(2,5-difluorophenyl)butanamide
2-Bromo-N-(2,5-difluorophenyl)butanamide

Hazard: Irritant

Formula: C₁₀H₁₀BrF₂NO
MDL Number: MFCD12027366
CAS Number: 1119452-44-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR