2-Bromo-N-(2,5-dimethoxyphenyl)propanamide

Supplier: Matrix Scientific

038475-500MG
101845-580EA 87.4 USD
101845-580
2-Bromo-N-(2,5-dimethoxyphenyl)propanamide
2-Bromo-N-(2,5-dimethoxyphenyl)propanamide

Hazard: Irritant

Formula: C₁₁H₁₄BrNO₃
MDL Number: MFCD01311977
CAS Number: 3245-91-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR