2-Bromo-N-(2,3-dimethylphenyl)butanamide

Supplier: Matrix Scientific

038468-500MG
101845-566EA 87.4 USD
101845-566
2-Bromo-N-(2,3-dimethylphenyl)butanamide
2-Bromo-N-(2,3-dimethylphenyl)butanamide

Hazard: Irritant

Formula: C₁₂H₁₆BrNO
MDL Number: MFCD09269398
CAS Number: 924969-58-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR