[(6-Ethoxy-3,4-dihydrospiro[chromene-2,1'-cyclohexan]-4-yl)thio]acetic acid

Supplier: Matrix Scientific

038453-500MG
101845-536EA 144.9 USD
101845-536
[(6-Ethoxy-3,4-dihydrospiro[chromene-2,1'-cyclohexan]-4-yl)thio]acetic acid
[(6-Ethoxy-3,4-dihydrospiro[chromene-2,1'-cyclohexan]-4-yl)thio]acetic acid

Hazard: Irritant

Formula: C₁₈H₂₄O₄S
MDL Number: MFCD12027341
CAS Number: 899710-30-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR