(2-Methyl-3-oxo-2,3-dihydro-4H-1,4-benzoxazin-4-yl)acetic acid

Supplier: Matrix Scientific

038443-500MG
101845-516EA 217.35 USD
101845-516
(2-Methyl-3-oxo-2,3-dihydro-4H-1,4-benzoxazin-4-yl)acetic acid
(2-Methyl-3-oxo-2,3-dihydro-4H-1,4-benzoxazin-4-yl)acetic acid

Hazard: Irritant

Formula: C₁₁H₁₁NO₄
MDL Number: MFCD08444407
CAS Number: 899710-24-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR