6-Chloro-2-ethyl-3,4-dihydro-2H-1,4-benzoxazine

Supplier: Matrix Scientific

038442-500MG
101845-514EA 332.35 USD
101845-514
6-Chloro-2-ethyl-3,4-dihydro-2H-1,4-benzoxazine
6-Chloro-2-ethyl-3,4-dihydro-2H-1,4-benzoxazine

Hazard: Irritant

Formula: C₁₀H₁₂ClNO
MDL Number: MFCD11603431
CAS Number: 890045-10-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR