N,N-Diethyl-N'-[(1-methyl-1,2,3,4-tetrahydroquinolin-6-yl)methyl]propane-1,3-diamine

Supplier: Matrix Scientific

038441-500MG
101845-512EA 217.35 USD
101845-512
N,N-Diethyl-N'-[(1-methyl-1,2,3,4-tetrahydroquinolin-6-yl)methyl]propane-1,3-diamine
N,N-Diethyl-N'-[(1-methyl-1,2,3,4-tetrahydroquinolin-6-yl)methyl]propane-1,3-diamine

Hazard: Irritant

Formula: C₁₈H₃₁N₃
MDL Number: MFCD12027334
CAS Number: 1119453-04-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR