N-[(1-Methyl-1,2,3,4-tetrahydroquinolin-6-yl)methyl]-3-morpholin-4-ylpropan-1-amine

Supplier: Matrix Scientific

038440-500MG
101845-510EA 162.15 USD
101845-510
N-[(1-Methyl-1,2,3,4-tetrahydroquinolin-6-yl)methyl]-3-morpholin-4-ylpropan-1-amine
N-[(1-Methyl-1,2,3,4-tetrahydroquinolin-6-yl)methyl]-3-morpholin-4-ylpropan-1-amine

Hazard: Irritant

Formula: C₁₈H₂₉N₃O
MDL Number: MFCD12027333
CAS Number: 1119450-30-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR