N-(2-Methylcyclohexyl)-N-[(1-methyl-1,2,3,4-tetrahydroquinolin-6-yl)methyl]amine

Supplier: Matrix Scientific

038437-500MG
101845-504EA 217.35 USD
101845-504
N-(2-Methylcyclohexyl)-N-[(1-methyl-1,2,3,4-tetrahydroquinolin-6-yl)methyl]amine
N-(2-Methylcyclohexyl)-N-[(1-methyl-1,2,3,4-tetrahydroquinolin-6-yl)methyl]amine

Hazard: Irritant

Formula: C₁₈H₂₈N₂
MDL Number: MFCD12027331
CAS Number: 1119450-29-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR