N-[(1-Butyl-1,2,3,4-tetrahydroquinolin-6-yl)methyl]-3-methoxypropan-1-amine

Supplier: Matrix Scientific

038436-500MG
101845-502EA 217.35 USD
101845-502
N-[(1-Butyl-1,2,3,4-tetrahydroquinolin-6-yl)methyl]-3-methoxypropan-1-amine
N-[(1-Butyl-1,2,3,4-tetrahydroquinolin-6-yl)methyl]-3-methoxypropan-1-amine

Hazard: Irritant

Formula: C₁₈H₃₀N₂O
MDL Number: MFCD12027330
CAS Number: 1119451-46-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR