(3-Ethoxypropyl)[(1-propyl-1,2,3,4-tetrahydroquinolin-6-yl)methyl]amine

Supplier: Matrix Scientific

038433-500MG
101845-496EA 162.15 USD
101845-496
(3-Ethoxypropyl)[(1-propyl-1,2,3,4-tetrahydroquinolin-6-yl)methyl]amine
(3-Ethoxypropyl)[(1-propyl-1,2,3,4-tetrahydroquinolin-6-yl)methyl]amine

Hazard: Irritant

Formula: C₁₈H₃₀N₂O
MDL Number: MFCD12027327
CAS Number: 1119451-45-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR