N-(2-Furylmethyl)-N-(2-methylcyclohexyl)amine

Supplier: Matrix Scientific

038430-500MG
101845-490EA 217.35 USD
101845-490
N-(2-Furylmethyl)-N-(2-methylcyclohexyl)amine
N-(2-Furylmethyl)-N-(2-methylcyclohexyl)amine

Hazard: Irritant

Formula: C₁₂H₁₉NO
MDL Number: MFCD01135365
CAS Number: 355381-97-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR