N-(3-Ethoxypropyl)-N-(2-furylmethyl)amine

Supplier: Matrix Scientific

038423-500MG
101845-476EA 144.9 USD
101845-476
N-(3-Ethoxypropyl)-N-(2-furylmethyl)amine
N-(3-Ethoxypropyl)-N-(2-furylmethyl)amine

Hazard: Irritant

Formula: C₁₀H₁₇NO₂
MDL Number: MFCD07410479
CAS Number: 892582-02-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR