5-{[(3-Methylbenzyl)sulfonyl]methyl}-2-furoic acid

Supplier: Matrix Scientific

038417-500MG
101845-462EA 217.35 USD
101845-462
5-{[(3-Methylbenzyl)sulfonyl]methyl}-2-furoic acid
5-{[(3-Methylbenzyl)sulfonyl]methyl}-2-furoic acid

Hazard: Irritant

Formula: C₁₄H₁₄O₅S
MDL Number: MFCD03834537
CAS Number: 696648-31-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR