5-{[(3-Methylbenzyl)thio]methyl}-2-furoic acid

Supplier: Matrix Scientific

038410-500MG
101845-448EA 217.35 USD
101845-448
5-{[(3-Methylbenzyl)thio]methyl}-2-furoic acid
5-{[(3-Methylbenzyl)thio]methyl}-2-furoic acid

Hazard: Irritant

Formula: C₁₄H₁₄O₃S
MDL Number: MFCD03834531
CAS Number: 696647-99-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR