N-(1-Thien-2-ylethyl)cyclohexanamine

Supplier: Matrix Scientific

038408-500MG
101845-444EA 217.35 USD
101845-444
N-(1-Thien-2-ylethyl)cyclohexanamine
N-(1-Thien-2-ylethyl)cyclohexanamine

Hazard: Irritant

Formula: C₁₂H₁₉NS
MDL Number: MFCD11138591
CAS Number: 1019474-95-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR