1-(4-Methylbenzyl)-5-oxopyrrolidine-3-carboxylic acid

Supplier: Matrix Scientific

038404-500MG
101845-436EA 217.35 USD
101845-436
1-(4-Methylbenzyl)-5-oxopyrrolidine-3-carboxylic acid
1-(4-Methylbenzyl)-5-oxopyrrolidine-3-carboxylic acid

Hazard: Irritant

Formula: C₁₃H₁₅NO₃
MDL Number: MFCD01934736
CAS Number: 96449-91-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR