N,2,3-Trimethylcyclohexanamine

Supplier: Matrix Scientific

038400-500MG
101845-428EA 290.77 USD
101845-428
N,2,3-Trimethylcyclohexanamine
N,2,3-Trimethylcyclohexanamine

Hazard: Irritant

Formula: C₉H₁₉N
MDL Number: MFCD11154125
CAS Number: 42195-94-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR