4-{[(4-Bromobenzyl)amino]methyl}benzoic acid

Supplier: Matrix Scientific

038397-500MG
101845-422EA 217.35 USD
101845-422
4-{[(4-Bromobenzyl)amino]methyl}benzoic acid
4-{[(4-Bromobenzyl)amino]methyl}benzoic acid

Hazard: Irritant

Formula: C₁₅H₁₄BrNO₂
MDL Number: MFCD03450460
CAS Number: 656815-39-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR